missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Crystal Violet lactone

Catalog No. p-7058976
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4371006 5 g
AA4371014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4371006 Supplier Thermo Scientific Chemicals Supplier No. 04371006

CAS: 1552-42-7 | C26H29N3O2 | 415.54 g/mol

Crystal Violet lactone is an important functional dye for the production of pressure sensitive materials. It is a chemical intermediate , intermediates for thermal/carbonless Paper, used in medicine and biochemical research, a leuco dye..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1552-42-7
Molecular Formula C26H29N3O2
Molecular Weight (g/mol) 415.54
MDL Number MFCD00070611
InChI Key IPAJDLMMTVZVPP-UHFFFAOYSA-N
PubChem CID 73773
IUPAC Name 6-(dimethylamino)-3,3-bis[4-(dimethylamino)phenyl]-2-benzofuran-1-one
SMILES CN(C)C1=CC=C(C=C1)C1(OC(=O)C2=CC(=CC=C12)N(C)C)C1=CC=C(C=C1)N(C)C

Specifications

Melting Point 180°C to 183°C
Solubility Information Insoluble in water.
Formula Weight 415.53
Quantity 5 g
Physical Form Powder
Chemical Name or Material Crystal Violet lactone
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 216-293-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only