missing translation for 'onlineSavingsMsg'
Learn More

Cryptotanshinone 97.0+%, TCI America™

Catalog No. C336325MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C336325MG 25 mg
C3363100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C336325MG Supplier TCI America Supplier No. C336325MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35825-57-1
Molecular Formula C19H20O3
Molecular Weight (g/mol) 296.366
MDL Number MFCD07636810
InChI Key GVKKJJOMQCNPGB-JTQLQIEISA-N
Synonym Tanshinone C, (R)-1,2,6,7,8,9-Hexahydro-1,6,6-trimethylphenanthro[1,2-b]furan-10,11-dione
PubChem CID 160254
IUPAC Name (1R)-1,6,6-trimethyl-2,7,8,9-tetrahydro-1H-naphtho[1,2-g][1]benzofuran-10,11-dione
SMILES CC1COC2=C1C(=O)C(=O)C3=C2C=CC4=C3CCCC4(C)C

Specifications

Melting Point 192°C
Quantity 25 mg
UN Number 1544
Formula Weight 296.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cryptotanshinone
TSCA No
Recommended Storage Refrigerator