missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryoware Permanent Markers and Pens

Catalog No. 1338288
Change view
Click to view available options
Color:
Assorted (Red, Blue, Green, Black)
Black
No. per Case:
10
4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Case
1338288 Black 4
12565237 Black 10
1338252 Assorted (Red, Blue, Green, Black) -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 1338288 Supplier Thermo Scientific™ Supplier No. 63130020

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Identify samples and storage boxes permanently with these cryoware permanent markers and pens.

Permanently notate samples and storage boxes with Thermo Scientific™ Cryoware Permanent Markers and Pens, which can easily withstand cold storage temperatures.

  • Permanent marker set of four
  • Extra-fine point
  • Ink color: Black

Specifications

Product Type Permanent Marking Pen
Tip Style Extra Fine
Color Black
Description Cryoware Permanent Marker
Includes Consists of four extra-fine-point pens
For Use With (Application) Used for marking cardboard and Polycarbonate
No. per Case 4
Quantity 1 Pk.