missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ CryoStorage Vials, 3D SafeCode

Catalog No. 05413992
Change view
Click to view available options
Volume (Metric):
0.5 mL
1.0 mL
1.5 mL
2.0 mL
4.0 mL
Sterility:
Non-sterile
Sterile
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility
05413992 4.0 mL Sterile
05413983 0.5 mL Non-sterile
05413984 1.0 mL Non-sterile
05413985 1.5 mL Non-sterile
05413986 2.0 mL Non-sterile
05413987 4.0 mL Non-sterile
05413988 0.5 mL Sterile
05413989 1.0 mL Sterile
05413990 1.5 mL Sterile
05413991 2.0 mL Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 05413992 Supplier Eppendorf™ Supplier No. 0030079493

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labeled vials are pre-racked and pre-capped (0.5 mL to 4.0 mL) for storage applications.

  • 3-level coding for safe sample identification
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -196 °C (gas phase of LN2) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Additional information on vials available via Eppendorf website for documentation purposes
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling

Specifications

Material Polypropylene
Sterility Sterile
Volume (Metric) 4.0 mL
No. per Pack 48
RNase Free Yes
Dnase Free Yes
Barcode 2D SafeCode
Quantity 480 (10 Racks of 48 Vials)
Product Type Coded Cryobank Vial System