Learn More
Stemcell Technologies CRYOSTOR CSB, SIZE 100 ML DFS Item
Supplier:  Stemcell Technologies Inc 1000237
CryoStor CSB is an intracellular-like base formulation from the CryoStor preservation media series. It is serum-free, animal component-free, and does not contain dimethyl sulfoxide (DMSO). It is designed to improve post-thaw cell viability and function, and to reduce the level of cryopreservation-induced delayed-onset cell death.
Catalog No. NC1861475
Only null left
Explore available promotions