Learn More
Stemcell Technologies CryoStor CS10, Size: 50 mL DFS Item
Supplier:  Stemcell Technologies Inc 07959
Maximize post-thaw cell recovery and viability following cryopreservation at very low temperatures (-80C to -196C) with ready-to-use CryoStor CS10 medium. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment that is recommended for the cryopreservation of a variety of sensitive cell and sample types, including myeloma cell lines, human pluripotent stem cells, human peripheral blood, and more. Available in a variety of convenient formats, CryoStor CS10 is formulated with USP-grade components to minimize variability and contains 10% DMSO. Visit STEMCELL.com/cryostor-cs10 for more information.
Numéro de catalogue. NC1530512
Il en reste null
Découvrez plus d'offres spéciales