missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CryoStar™ NX70 Cryostat with Cold D and Vacutome, High-Profile Blade Carrier

Accommodate the needs of both research and clinical laboratories with the Thermo Scientific™ CryoStar™ NX70.

Supplier:  Thermo Scientific™ 957030

Catalog No. 36-102-8254

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Accommodate the needs of both research and clinical laboratories with the the Thermo Scientific™ CryoStar™ NX70. The NX70 introduces a form-fitting design, integrated height control and motorized sectioning.

Standard Features
  • Knife advance
  • Designations for both low and high blade holders
  • Multi-axis specimen orientation
  • High quality motorized sectioning
  • Specimen retraction
Optional Features
  • Cold Disinfection with Cryostar NX70 D
  • Vacutome with Cryostar NV70 V
  • Both Cold Disinfection and Vacutome with Cryostar NX70 DV
Ease of Use
  • Large open chamber with bright LED lighting
  • Integrated color LCD touch screen
  • Joystick
Temperature Control
  • Precise control through periods of prolonged use 
  • Rapid Response Temperature Control cools object holder, blade holder and cryobar only
Comfortable Ergonomic Design
  • Integrated electronic hight adjustment from 32 to 44 inches
  • Form-fitting front panel and arm rest to reduce user fatigue
Includes: Low-profile blade holder, high-profile blade holder or 16cm steel knife carrier
 
Ordering Information: Low- or High-profile blade holder or 16cm steel knife carrier must be designated at time of ordering.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions