missing translation for 'onlineSavingsMsg'
Learn More

Ad Infinitum™ CryoProtectPURE STEM 5X™ DMSO-Free/Serum-Free Cryopreservation Media

Catalog No. CPP300025
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CPP300025 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CPP300025 Supplier Ad Infinitum™ Supplier No. CPP300025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CryoProtectPURE STEM 5X™ DMSO-Free/Serum-Free Cryopreservation Media is optimized for stem cell and cord blood applications. Premixed, ready-to-use.

CryoProtectPURE STEM 5X DMSO-Free/Serum-Free Cryopreservation Media increases cell viability, produces healthier cells post-thaw with no antigenicity concern or toxic vitrification, produces higher repeatability, higher viability, and increased preservation of expression post thaw.
  • DMSO-Free
  • Serum-Free
  • Pre-mixed, ready-to-use, 0.2 μm filtered.
  • Optimized for Cord Blood and Stem Cell Applications.
  • Shipped at ambient temperature.
  • Store at 2 to 8°C.
  • For manufacturing and/or research use only, not for diagnostic or therapeutic use.
  • Manufactured at a cGMP-compliant facility registered with the FDA as a medical device manufacturer and certified to ISO 13485 standards.
  • Specifications

    Quantity 25 mL
    Content And Storage 2°C to 8°C

    For manufacturing and/or research use only, not for diagnostic or therapeutic use.