missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Cryopreserved Human Kupffer Cells

Catalog No. HUKCCS
Encompass
Change view
Click to view available options
Quantity:
1 vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
HUKCCS 1 vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. HUKCCS Supplier Gibco™ Supplier No. HUKCCS
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cryopreserved Human Kupffer Cells

Cryopreserved Human Kupffer Cells are isolated from adult human livers. Each vial contains a minimum of one million viable cells. These easy-to-use cells provide a convenient way to produce hepatocyte and Kupffer cell co-cultures for the study of various hepatic functions.These cells typically have high viabilities and in vivo-like expression levels.

  • Viability routinely >90%
  • Purity routinely >90%
  • Respond to activation with lipopolysaccharides (LPS)
  • Minimum 1 million viable cells per vial
  • Protocols for hepatocyte co-culture available

    Kupffer Cells Provide for Improved Physiological Modeling
    Growing evidence shows that under both normal and pathological conditions, many hepatocyte functions are regulated by substances released from neighboring non-parenchymal cells (NPC). These cells, particularly Kupffer cells, play an important role in the modulation of xenobiotic metabolism in the liver. Kupffer cells secrete potent mediators of the inflammatory response that controls liver inflammation. These cytokine mediators control hepatocyte metabolic rates through direct interactions with phase I and phase II enzymes. Kupffer cell and hepatocyte co-cultures can self-assemble within 72 hours of treatment with pro-inflammatory cytokines or LPS, and can function by effectively modulating P450 expression, thus giving a more physiologically relevant result. Studies indicate that co-culture of hepatocytes with NPCs better represent both normal liver physiology as well as disease states.
  • Specifications

    Content And Storage 1 vial of 1 million viable human Kupffer cells.

    Store in a cryostorage dewar or -135°C freezer.
    Cell Type Macrophage Cells
    Culture Type Adherent Cell Culture
    Form Cryopreserved
    Species Human
    Donor Source Single Donor
    No. of Cells 1 x 106
    Product Type Human Kupffer Cell
    Quantity 1 vial

    For Research Use Only. Not for use in diagnostic procedures.