Learn More
Thermo Scientific™ CryoPlus™ Series Racks and Holders
Increase the usability of Thermo Scientific™ CryoPlus™ LN2 storage systems with CryoPlus Series Racks and Holders.
Supplier:  Thermo Scientific™ 4000011
 View more versions of this product
Catalog No. 15466189
Only null left
Description
Description
Increase the usability of Thermo Scientific™ CryoPlus™ LN2 storage systems with CryoPlus Series Racks and Holders.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
Specifications
Specifications
Vertical Rack, Vapor Phase
4 Boxes
4 Boxes
CryoPlus 1,2,3,4
Vials
Product Suggestions
Product Suggestions