missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cryogenic Vial Gripper
GREENER_CHOICE

Catalog No. 17100481
Change view
Click to view available options
For Use With (Equipment):
Cryogenic Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
17100481 Cryogenic Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17100481 Supplier Fisherbrand™ Supplier No. 120908F2

The perfect Cryogenic grip for optimal precision

  • These two styles of hand-operated Grippers are designed to pick up cryogenic and crimped 10R-size vaccine/serum vials, respectively
  • Both are durably made of reinforced polypropylene which is cold-tolerant and resistant to everyday wear
  • The designated squeeze target ensures maximum precision and grip with every use
  • Each style is available in a pack of 5 assorted colors for easy identification and organization
  • A combo pack with one of each style is also available

Specifications

Type Vial Gripper
Color Assorted (Red, Blue, Green, Yellow, Purple)
For Use With (Equipment) Cryogenic Vials