Learn More
GA International Cryogenic Laser Labels for Frozen Vials and Containers 0.94"" x 0.5"" / 23.9mm x 12.7mm Rectangle Orange From -196C to +100C (-321F to +212F)
Catalog No. 5000572273
Encompass
These cryogenic labels are designed to be applied directly, and will rapidly adhere, to frozen cryo vials, tubes, cryo boxes, as well as plastic and cardboard freezer boxes, which are difficult to label under deep-freeze conditions (as low as -80C/-112F). Designed for long-term preservation in liquid nitrogen (-196C/-321F), -80C (-112F) freezers, for dry ice transportation, and can withstand temperatures up to +100C (+212F)*.Printable with all common brands of laser printers to obtain a sharp, smudge-proof printout that is also resistant to mild laboratory chemicals (spray and swab). They can be applied on frozen vials and tubes without the need of an overlap and can also be written on with our Cryo-Markers.*These labels have been tested to withstand up to 2 hours of 150C/302F dry heat. Some softening of the adhesive is observed, which is restored when the label is returned to ambient conditions.
Catalog No. 5000572273 Supplier GA International Supplier No. LCS12OR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These cryogenic labels are designed to be applied directly, and will rapidly adhere, to frozen cryo vials, tubes, cryo boxes, as well as plastic and cardboard freezer boxes, which are difficult to label under deep-freeze conditions (as low as -80C/-112F). Designed for long-term preservation in liquid nitrogen (-196C/-321F), -80C (-112F) freezers, for dry ice transportation, and can withstand temperatures up to +100C (+212F)*.Printable with all common brands of laser printers to obtain a sharp, smudge-proof printout that is also resistant to mild laboratory chemicals (spray and swab). They can be applied on frozen vials and tubes without the need of an overlap and can also be written on with our Cryo-Markers.*These labels have been tested to withstand up to 2 hours of 150C/302F dry heat. Some softening of the adhesive is observed, which is restored when the label is returned to ambient conditions.