These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for liquid nitrogen storage (Subject to prior verification of compatibility of labels and intended vial cap material in Liquid Nitrogen), -80C freezers and dry ice. Specially designed to comfortably fit cryovials and Eppendorf tube caps. These waterproof deep-freeze dot labels are supplied in roll format and are available in 15 individual colors as well as packs of 15 assorted colors for color-coding and rapid identification.