missing translation for 'onlineSavingsMsg'
Learn More

National Safety Apparel™ Cryogenic Apron

Catalog No. 17355082
Change view
Click to view available options
Dimensions (L x W):
24 x 36 in.
24 x 42 in.
24 x 48 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17355082 24 x 42 in.
17355081 24 x 36 in.
17355083 24 x 48 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17355082 Supplier National Safety Apparel™ Supplier No. A02CRC24X42
Only null left

Cryogenic apron provides waterproof, thermal protection against liquid nitrogen splashes and other extremely cold temperature tasks.

  • Constructed of Teflon™ Laminated Nylon
  • 3M™ Scotchlite™ Thinsulate™ liner
  • Water resistant
  • Adjustable strap and buckle closure at waist and neck
  • Made in the USA

Specifications

Type Apron
Size Universal
Material Nylon
Color Blue
Closure Type Buckles (Rear)
Sterility Sterile
Finish Laminated
Latex Free Yes
Neck Style Adjustable
For Use With (Application) Cryogenic Liquids, Gases
Certifications/Compliance EN 511 Contact Cold Performance Level 2, EN 511 Convective Cold Performance Level 3
Dimensions (L x W) 24 x 42 in.
Length (English) 42 in.
Width (English) 24 in.
Show More Show Less