Learn More
Thermo Scientific™ CryoBox™ Freezer Racks
Stack and organize standard cryogenic vial storage boxes with these freezer racks.
Supplier: Thermo Scientific™ DS50350009
Description
Stack and organize standard cryogenic vial storage boxes with these freezer racks. Thermo Scientific™ CryoBox™ Freezer Racks include durable Thermo Scientific Nalgene™ Stainless Steel CryoBox Racks and Thermo Scientific Nunc™ aluminum racks that fit most common upright and chest freezers.
- Conveniently store standard 5-1/4 x 5-1/4 cryogenic vial storage boxes with these freezer racks
- Fits Thermo Scientific Nalgene™ and Nunc™ CryoBoxes™
- Options include durable stainless-steel and aluminum racks that fit most common upright and chest freezers
Chemical Identifiers
| Silver | |
| Stainless Steel |
Specifications
| Silver | |
| 9 CryoBoxes/Rack | |
| Stainless Steel |
| 8.4 x 8.3 x 50.2 cm | |
| Vertical | |
| Freezer Rack |