missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Cryo Storage Box

Catalog No. S15786
Change view
Click to view available options
Holds:
1.0 and 1.8 mL Vials
4.5 mL Vials
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
S15786 1.0 and 1.8 mL Vials
S15914 4.5 mL Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S15786 Supplier United Scientific™ Supplier No. P20602
Only null left

Safely store your cryogenic samples.

  • Pack of four autoclavable polycarbonate boxes, each designed for at temperatures as low as -196°C.
  • Available in two variations, both hold 81 vials in a 9 x 9 Array. Small size fits 1 and 1.8 mL vials and large size fits 4.5 mL vials.
  • Compatible with both internal and external threaded cryogenic vials.
  • These boxes provide a small, space-efficient storage system within a freezer.
  • The transparent lid features a numbered grid for easy identification of contents.
  • Not recommended for use in the liquid phase of liquid nitrogen.
  • Can be used in vapor phase of liquid nitrogen.

Specifications

Color White
Material Polycarbonate
Shape Square
Type Cryo Storage Box
Holds 1.0 and 1.8 mL Vials