missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Marker Pen Set

Catalog No. 11679117
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11679117 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11679117 Supplier Thermo Scientific™ Supplier No. 4000222

Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.

Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.

  • Set includes 4 black pens.

Specifications

Type Cryo Marker Pen Set
Temperature Range -200°C, -328°F
Color Black
For Use With (Equipment) cryovials and boxes
Includes 4 black pens