Learn More
GA International Cryogenic Lamination Tape 2"" x 200' / 50.8mm x 60m Transparent From -196C to +93C (-321F to +199F)
Catalog No. NC0831007
Encompass
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as -196C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.
Catalog No. NC0831007 Supplier GA International Supplier No. CLAM50C3200
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as -196C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.