Learn More
Alkali Scientific ASI™ Cryo Label Sheets, 38x19mm for General Use, Red, 1200 labels, 20 sheets/Cs
Catalog No. 503027718
Encompass
CRYO LABEL SHEETS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194 F (90C) to as low as -94F (-70C).
Catalog No. 503027718 Supplier Alkali Scientific Supplier No. LCL34112SR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CRYO LABEL SHEETS 38x19mm for General Use We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194 F (90C) to as low as -94F (-70C).