Alkali Scientific ASI™ Cryo Label Rolls, 33x13mm for 1.5/2mL Tubes, 1000 per Roll, 1/Roll, Orange
Catalog No. 503027618
CRYO LABEL ROLLS 33x13mm for 1.5/2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194 F (90C) to as low as -94F (-70C). Labels Per roll: 1000, 1 roll/cs
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CRYO LABEL ROLLS 33x13mm for 1.5/2ml Tubes We offer cryogenic storage labels that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194 F (90C) to as low as -94F (-70C). Labels Per roll: 1000, 1 roll/cs