Learn More
Tempshield™ Cryo-Gloves™ Pink Mid-Arm Length Cryogenic Gloves
State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).
$406.71 - $482.56
Specifications
Certifications/Compliance (Europe) EN 420, EN 511
Description Mid-Arm Length
Color Pink
Hand Specific
Product Type Cryogenic Gloves
View More Specs
Alert:
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-230-261
View Documents
Tempshield™
PMAS
Small
Pair for $482.56
Case of 10 Pair for $4,387.00
Only null left
19-230-262
View Documents
Tempshield™
PMAM
Medium
Pair for $482.56
Case of 10 Pair for $4,387.00
Only null left
19-230-263
View Documents
Tempshield™
PMAL
Large
Pair for $406.71
Case of 10 Pair for $3,697.40
Only null left
19-230-264
View Documents
Tempshield™
PMAXL
X-Large
Pair for $406.71
Case of 10 Pair for $3,697.40
Only null left
Description
Description
State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).
  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids
Specifications
Specifications
EN 420, EN 511
Pink
Cryogenic Gloves
Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
-320°F
13.3 to 15.5 in.
Mid-Arm Length
Specific
Yes
Cryo-Gloves™
-196°C
33.5 to 39.5 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'