missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryo Gloves - Elbow Length

Catalog No. p-7219199
Change view
Click to view available options
Size:
Medium
Large
Catalog No. Size
15466317 Medium
15466319 Large
2 options

Catalog No. 15466317

Supplier: Thermo Scientific™ 189564

Only null left

Safely handle samples from ultra-low and cryogenic storage with these cryo gloves which offer a high level of thermal protection.

  • Comfortable, lightweight, and breathable protection
  • Side release buckles for quick on/off
  • Available in a variety of sizes

Specifications

Description Elbow Length
Color Blue
Hand Specific
Size Medium
For Use With (Application) Handling Samples from Ultra Low Temperature and Cryogenic Storage
Length (English) 18 in.
Length (Metric) 45.7 cm