missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Waterproof Mid-Arm Length Cryogenic Gloves

Catalog No. p-7219189
Encompass
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11394325 Small
11394326 Medium
11394327 Large
11394328 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Catalog No. 11394325 Supplier Tempshield™ Supplier No. MASWP
Only null left
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

A thin, seamless 100% waterproof liner provides protection from spills and splashes. State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • Thin, seamless 100% waterproof liner provides protection from spills and splashes
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Mid-Arm Length
Color Blue
Hand Specific
Product Type Waterproof Cryogenic Gloves
Size Small
EN388 Standard Yes
For Use With (Application) Dispensing/Transferring Cryogenic Liquids, Removing Samples from Cryogenic Liquids, and Open Cryogenic Containers with the Chance of Exposure to Liquids/Splashes
Series Cryo-Gloves™
Temperature (English) Min -320°F
Temperature (Metric) Min. -196°C
Length (English) 13.3 to 15.3 in.
Length (Metric) 33.5 to 39.5 cm
Show More Show Less