missing translation for 'onlineSavingsMsg'
Learn More

Tempshield™ Cryo-Gloves™ Blue Mid-Arm Length Cryogenic Gloves

Catalog No. p-7219195
Encompass
Change view
Click to view available options
Size:
Medium
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11394305 Medium
11394200 Large
19059925 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.
Catalog No. 11394305 Supplier Tempshield™ Supplier No. MAM
Only null left
Not for immersion in liquid nitrogen, for use near open flame, or in hazardous areas where ignition could occur.

State-of-the art materials and multi-layered construction provide cryogenic protection for ultra-cold applications down to -196°C (-320°F).

State-of-the art materials and multi-layered construction provides cryogenic protection for ultra-cold applications down to -196°C (-320°F).

  • Cryogenic protection for ultra-cold applications down to -196°C (-320°F)
  • State-of-the art materials and multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity
  • High performance, thermal lining wicks moisture away from hands to maintain comfort over extended periods of wear
  • Ideal for applications where function is important and safety is critical
  • Not intended for immersion in liquid nitrogen or other cryogenic liquids

Specifications

Certifications/Compliance (Europe) EN 420, EN 511
Description Mid-Arm Length
Color Blue
Hand Specific
Product Type Cryogenic Gloves
Size Medium
EN388 Standard Yes
For Use With (Application) Dry Ice Handling, Low/Ultra-Low Temperature Freezers, and Closed Cryogenic Systems
Series Cryo-Gloves™
Temperature (English) Min -320°F
Temperature (Metric) Min. -196°C
Length (English) 13.3 to 15.5 in.
Length (Metric) 33.5 to 39.5 cm
Show More Show Less