Learn More
MiTeGen Llc CX100 Dry Shipper by IC Biomedical & Square Shipping Case Bundle, Ideal For -190C Cryogenic Transport, IATA Compliant
Catalog No. NC1551159
Encompass
This bundle features the CX100 Cryo Express Dry Shipper by IC Biomedical paired with a robust square shipping case, providing a complete solution for secure cryogenic material transport. The CX100 Dry Shipper is engineered to safely move various materials at approximately -190C. Its unique adsorbent material prevents liquid nitrogen spills even if tipped, ensuring reliable transit and IATA compliance for non-infectious biological samples. The included square shipping case, measuring 17" x 17" x 25" and weighing 32 lbs, offers superior physical protection. Made of thick, hard plastic with a padded lining, it features a secure latching mechanism and side handles for easy, safe transport. The CX100 offers an 18-day static holding time and 4.1 liters LN2 adsorbed capacity.
Catalog No. NC1551159 Supplier MiTeGen Llc Supplier No. TWCXS1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This bundle features the CX100 Cryo Express Dry Shipper by IC Biomedical paired with a robust square shipping case, providing a complete solution for secure cryogenic material transport. The CX100 Dry Shipper is engineered to safely move various materials at approximately -190C. Its unique adsorbent material prevents liquid nitrogen spills even if tipped, ensuring reliable transit and IATA compliance for non-infectious biological samples. The included square shipping case, measuring 17" x 17" x 25" and weighing 32 lbs, offers superior physical protection. Made of thick, hard plastic with a padded lining, it features a secure latching mechanism and side handles for easy, safe transport. The CX100 offers an 18-day static holding time and 4.1 liters LN2 adsorbed capacity.