Learn More
MiTeGen Llc Cryo-EM Puck Grasping Tongs, For Easy & Secure Handling of Cryogenic Storage Pucks, Enhances Workflow
Catalog No. NC1863749
Encompass
These Cryo-EM Puck Grasping Tongs are expertly designed for the safe, easy, and precise manipulation of cryo-EM storage pucks, such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1, in demanding laboratory environments. Engineered to provide exceptional control, these tongs allow users to handle pucks with enhanced security, minimizing direct contact with cryogenic surfaces and reducing the risk of thermal shock or accidental drops. Their specialized design ensures a firm and stable grip on storage pucks, streamlining transfers between liquid nitrogen (LN2) dewars, workstations, and automated systems. This tool is indispensable for maintaining sample integrity and optimizing workflow efficiency in cryo-electron microscopy applications. By facilitating quick and secure handling, these grasping tongs contribute to more reliable experimental setups and a safer working environment for valuable biological samples stored at ultra-low temperatures.
Catalog No. NC1863749 Supplier MiTeGen Llc Supplier No. MCEMG2PGT1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
These Cryo-EM Puck Grasping Tongs are expertly designed for the safe, easy, and precise manipulation of cryo-EM storage pucks, such as the MiTeGen Generation 2 cryoem puck SKU M-CEMG2-1, in demanding laboratory environments. Engineered to provide exceptional control, these tongs allow users to handle pucks with enhanced security, minimizing direct contact with cryogenic surfaces and reducing the risk of thermal shock or accidental drops. Their specialized design ensures a firm and stable grip on storage pucks, streamlining transfers between liquid nitrogen (LN2) dewars, workstations, and automated systems. This tool is indispensable for maintaining sample integrity and optimizing workflow efficiency in cryo-electron microscopy applications. By facilitating quick and secure handling, these grasping tongs contribute to more reliable experimental setups and a safer working environment for valuable biological samples stored at ultra-low temperatures.