missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ Crucible Sealing Press

Catalog No. 01915204
Change view
Click to view available options
Diameter (English):
6 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English)
01915204 6 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01915204 Supplier METTLER TOLEDO™ Supplier No. 119410

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The press allows the pan to be sealed very easily. Under the pressure of the plunger the pan is cold welded hermetically with the lid. After changing plunger and die you can use the press for other crucibles.

The press allows the pan to be sealed very easily. Under the pressure of the plunger the pan is cold welded hermetically with the lid. After changing plunger and die you can use the press for other crucibles.

For use with 30421563, 30421564, 30319250, 30319252, 30421562, 30319254, 30293533, 30319177, 30319178 & 30084906

Specifications

Product Type Sealing Press
Diameter (English) 6 in.
For Use With (Application) Crucibles
For Use With (Equipment) 30421563
Weight (English) 5.4 lb.