missing translation for 'onlineSavingsMsg'
Learn More

Crotyl Chloride (cis- and trans- mixture, contains 38% 3-Chloro-1-butene at maximum) 62.0+%, TCI America™

Catalog No. C1686500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1686500ML 500 mL
C168625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1686500ML Supplier TCI America Supplier No. C1686500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 591-97-9
Molecular Formula C4H7Cl
Molecular Weight (g/mol) 90.55
MDL Number MFCD00000987
InChI Key YTKRILODNOEEPX-NSCUHMNNSA-N
Synonym 1-chloro-2-butene, crotyl chloride, e-1-chlorobut-2-ene, 2-butenyl chloride, e-1-chloro-2-butene, 1-chlorobut-2-ene, krotylchlorid czech, gamma-methallyl chloride, gamma-methylallyl chloride, unii-xx3qn28jux
PubChem CID 637923
IUPAC Name (E)-1-chlorobut-2-ene
SMILES CC=CCCl

Specifications

Color Yellow
Quantity 500 mL
UN Number 1993
Formula Weight 90.55
Percent Purity ≥62.0% (GC)
Physical Form Liquid
Chemical Name or Material Crotyl Chloride (cis- and trans- mixture, contains 38% 3-Chloro-1-butene at maximum)
EINECSNumber (2)-0117
RTECSNumber EM4264000
TSCA Yes