missing translation for 'onlineSavingsMsg'
Learn More

Crotonic Anhydride 98.0+%, TCI America™

Catalog No. c042025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C042025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C042025G Supplier TCI America Supplier No. C042025G

Specifications

CAS 623-68-7
Color Yellow
Boiling Point 131°C
Molecular Formula C8H10O3
Quantity 25 g
MDL Number MFCD00009286
UN Number 3265
Synonym crotonic anhydride, 2-butenoic anhydride, 2-butenoic acid, anhydride, crotonic acid anhydride, but-2-enoic anhydride, unii-n521pmq949, anhydrid kyseliny krotonove czech, crotonicanhydride, e-but-2-enoyl e-but-2-enoate, anhydrid kyseliny krotonove
InChI Key VJDDQSBNUHLBTD-GGWOSOGESA-N
SMILES CC=CC(=O)OC(=O)C=CC
IUPAC Name [(E)-but-2-enoyl] (E)-but-2-enoate
Molecular Weight (g/mol) 154.165
PubChem CID 6049834
Formula Weight 154.17
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Crotonic Anhydride
Show More Show Less
RTECSNumber GQ6125000
TSCA Yes