missing translation for 'onlineSavingsMsg'
Learn More

Crotonic Acid 98.0+%, TCI America™

Catalog No. C041625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041625G 25 g
C0416100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C041625G Supplier TCI America Supplier No. C041625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-93-7
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00002701
InChI Key LDHQCZJRKDOVOX-NSCUHMNNSA-N
Synonym crotonic acid, e-but-2-enoic acid, but-2-enoic acid, trans-2-butenoic acid, trans-crotonic acid, 3-methylacrylic acid, e-2-butenoic acid, 2-butenoic acid, 2e-but-2-enoic acid, e-crotonic acid
PubChem CID 637090
ChEBI CHEBI:41131
IUPAC Name (E)-but-2-enoic acid
SMILES CC=CC(=O)O

Specifications

Melting Point 71°C
Color White-Yellow
Boiling Point 185°C
Quantity 25 g
UN Number 2823
Formula Weight 86.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Crotonic Acid
EINECSNumber (2)-0963
RTECSNumber GQ2800000
TSCA Yes