missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotonic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 71°C |
| Color | White-Yellow |
| Boiling Point | 185°C |
| Quantity | 100 g |
| UN Number | 2823 |
| Formula Weight | 86.09 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Crotonic Acid |
Safety and Handling
| EINECSNumber | (2)-0963 |
| RTECSNumber | GQ2800000 |
| TSCA | Yes |