missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Crotonamide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 158°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 85.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Crotonamide |
Safety and Handling
| EINECSNumber | (9)-0392 |
| RTECSNumber | GQ0700000 |
| TSCA | No |