missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich Crotonaldehyde, mixture of cis and trans
Description
Crotonaldehyde, mixture of cis and trans
Specifications
Specifications
| CAS | 4170-30-3 |
| Density | 0.853 g/mL (at 20°C (literature)) |
| Boiling Point | 101°C to 102°C (lit.) |
| Molecular Formula | C4H6O |
| Refractive Index | n20/D 1.437 |
| Linear Formula | CH3CH=CHCHO |
| MDL Number | MFCD00007003 |
| Quantity | 250 mL |
| Synonym | 2-Butenal, mixture of cis and trans |
| Molecular Weight (g/mol) | 70.09 |
| Show More |