missing translation for 'onlineSavingsMsg'
Learn More

Croton oil

Catalog No. AAJ6236709
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6236709 10 g
AAJ6236714 25 g
AAJ6236722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6236709 Supplier Thermo Scientific Chemicals Supplier No. J6236709
Only null left

A natural source of phorbol, phorbol esters and glycerides of saturated fatty acids | CAS: 8001-28-3

Croton Oil is a natural source of phorbol, phorbol esters and glycerides of saturated fatty acids. Croton seeds are sometimes applied directly to the skin for muscle and joint pain. The main use for croton oil today is as an active ingredient in phenol-based facial peels. Dermatologists, plastic surgeons and other practitioners can vary the amount of croton oil in the peel to customize the treatment for a particular patient or skin area, such as on the neck or around the eyes. In Malaysia, people use croton oil in lamps and in soap.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Croton Oil is a natural source of phorbol, phorbol esters and glycerides of saturated fatty acids. Croton seeds are sometimes applied directly to the skin for muscle and joint pain. The main use for croton oil today is as an active ingredient in phenol-based facial peels. Dermatologists, plastic surgeons and other practitioners can vary the amount of croton oil in the peel to customize the treatment for a particular patient or skin area, such as on the neck or around the eyes. In Malaysia, people use croton oil in lamps and in soap.

Solubility
Soluble in alcohol, chloroform, ether, and petroleum ether. Insoluble in water.

Notes
Supplied as a crude extract

Chemical Identifiers

CAS 8001-28-3
MDL Number MFCD00130880
Synonym Croton resin; Oleum tiglii

Specifications

Color Brown to Yellow
Flash Point 74°C (165°F)
Odor Pungent
Quantity 10 g
Solubility Information Soluble in alcohol,chloroform,ether,and petroleum ether. Insoluble in water.
Physical Form Oil
Chemical Name or Material Croton oil
Hazard Category H227-H315-H319-H350
Hazard Statement GHS H Statement
H350-H315-H319-H227
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P201-P202-P210-P264b-P280i-P281-P302+P352-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
RTECSNumber GQ6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only