Learn More
DWK Life Sciences Kimble™ Cross Stir Bar
This all-purpose stir bar permits stirring in closed containers without contamination. Ideal for use with conical bottom HPLC reservoirs.
Supplier:  DWK Life Sciences 7911900100
 View more versions of this product
Catalog No. K7911900100
Only null left
Description
Description
  • PTFE-covered X-shaped magnetic stirring bar
  • More turbulence with less vibration than straight stir bars
  • Autoclavable
Specifications
Specifications
Cross Stir Bar
Fits 2L Reservoir
Product Suggestions
Product Suggestions