Learn More
Croscarmellose Sodium, NF, Spectrum™ Chemical
C1366, 74811-65-7
$620.06 - $5187.97
Chemical Identifiers
CAS 74811-65-7
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18601879
View Documents
Spectrum Chemical Mfg Cor
C1366500GM
500 g Poly Bottle
Each for $620.06
Only null left
18601880
View Documents
Spectrum Chemical Mfg Cor
C136625KG
2.5 kg Poly Pail
Each for $1,777.57
Only null left
18601881
View Documents
Spectrum Chemical Mfg Cor
C136625KGBL
25 kg Fiber Drum
Each for $5,187.97
Only null left
Description
Description
Spectrum™ Chemical Croscarmellose Sodium, NF is used commonly as a FDA-approved disintegrant in pharmaceutical manufacturing. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Chemical Identifiers
74811-65-7
Specifications
74811-65-7
14.0-28%
Poly Bottle
NF
100%
10%
500 g
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'