missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cromolyn Disodium Salt Hydrate, TCI America™
Supplier: TCI America C252125G
Specifications
| Cromolyn Disodium Salt Hydrate | |
| White-Yellow | |
| MFCD00057744 | |
| cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal | |
| [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O | |
| 512.33 | |
| CHEBI:128458 | |
| ≥98.0% (HPLC,T) |
| 15826-37-6 | |
| C23H14Na2O11 | |
| 25 g | |
| VLARUOGDXDTHEH-UHFFFAOYSA-L | |
| disodium 5-{3-[(2-carboxylato-4-oxo-4H-chromen-5-yl)oxy]-2-hydroxypropoxy}-4-oxo-4H-chromene-2-carboxylate | |
| 27503 | |
| 512.33 | |
| Crystalline Powder |
Chemical Identifiers
| 15826-37-6 | |
| 512.33 | |
| VLARUOGDXDTHEH-UHFFFAOYSA-L | |
| 27503 | |
| disodium 5-{3-[(2-carboxylato-4-oxo-4H-chromen-5-yl)oxy]-2-hydroxypropoxy}-4-oxo-4H-chromene-2-carboxylate |
| C23H14Na2O11 | |
| MFCD00057744 | |
| cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal | |
| CHEBI:128458 | |
| [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O |
Safety and Handling
EINECSNumber : (9)-1037
RTECSNumber : DJ2380000
TSCA : No