missing translation for 'onlineSavingsMsg'
Learn More

Cromolyn Disodium Salt Hydrate, TCI America™

Catalog No. C25215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25215G 5 g
C252125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25215G Supplier TCI America Supplier No. C25215G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 15826-37-6
Molecular Formula C23H14Na2O11
Molecular Weight (g/mol) 512.33
MDL Number MFCD00057744
InChI Key VLARUOGDXDTHEH-UHFFFAOYSA-L
Synonym cromolyn sodium, sodium cromoglycate, disodium cromoglycate, sodium cromoglicate, sodium cromolyn, gastrocrom, nasalcrom, cromoglycate disodium, natrium cromoglicat, lomudal
PubChem CID 27503
ChEBI CHEBI:128458
IUPAC Name disodium 5-{3-[(2-carboxylato-4-oxo-4H-chromen-5-yl)oxy]-2-hydroxypropoxy}-4-oxo-4H-chromene-2-carboxylate
SMILES [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 512.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Cromolyn Disodium Salt Hydrate
EINECSNumber (9)-1037
RTECSNumber DJ2380000
TSCA No