missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Croconic Acid 98.0+%, TCI America™
$306.10 - $1041.52
Chemical Identifiers
| CAS | 488-86-8 |
|---|---|
| Molecular Formula | C5H2O5 |
| Molecular Weight (g/mol) | 142.066 |
| MDL Number | MFCD00181389 |
| InChI Key | RBSLJAJQOVYTRQ-UHFFFAOYSA-N |
| Synonym | croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # |
| PubChem CID | 546874 |
| IUPAC Name | 4,5-dihydroxycyclopent-4-ene-1,2,3-trione |
| SMILES | C1(=C(C(=O)C(=O)C1=O)O)O |
Chemical Identifiers
| 488-86-8 | |
| 142.066 | |
| RBSLJAJQOVYTRQ-UHFFFAOYSA-N | |
| 546874 | |
| C1(=C(C(=O)C(=O)C1=O)O)O |
| C5H2O5 | |
| MFCD00181389 | |
| croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # | |
| 4,5-dihydroxycyclopent-4-ene-1,2,3-trione |
Specifications
| 488-86-8 | |
| Yellow | |
| MFCD00181389 | |
| croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # | |
| C1(=C(C(=O)C(=O)C1=O)O)O | |
| 142.066 | |
| 142.07 | |
| Crystalline Powder |
| 114°C | |
| C5H2O5 | |
| 1 g | |
| RBSLJAJQOVYTRQ-UHFFFAOYSA-N | |
| 4,5-dihydroxycyclopent-4-ene-1,2,3-trione | |
| 546874 | |
| ≥98.0% (HPLC) | |
| Croconic Acid |
Safety and Handling
TSCA : No