Learn More
Croconic acid, 98%
CAS: 488-86-8 | C5H2O5 | 142.066 g/mol
$406.01 - $1465.21
Chemical Identifiers
| CAS | 488-86-8 |
|---|---|
| Molecular Formula | C5H2O5 |
| Molecular Weight (g/mol) | 142.066 |
| MDL Number | MFCD00181389 |
| InChI Key | RBSLJAJQOVYTRQ-UHFFFAOYSA-N |
| Synonym | croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # |
| PubChem CID | 546874 |
| IUPAC Name | 4,5-dihydroxycyclopent-4-ene-1,2,3-trione |
| SMILES | C1(=C(C(=O)C(=O)C1=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2180903
|
Thermo Scientific Chemicals
B2180903 |
1 g |
Each for $406.01
|
|
|||||
|
AAB2180906
|
Thermo Scientific Chemicals
B2180906 |
5 g |
Each for $1,465.21
|
|
|||||
Description
Croconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCroconic acid is used to prepare 1,3-bis-(2-dimethylamino-5-thienyl)croconine by reacting with dimethyl-thiophen-2-yl-amine. It is also used in organic synthesis and employed as a dye for biological research purposes. Further, it is involved in the preparation of ethers such as dimethyl croconate. In addition to this, it acts as a ligand and form coordination complexes with metals such as barium, copper, silver and lead.
Solubility
Soluble in water and ethanol.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Chemical Identifiers
| 488-86-8 | |
| 142.066 | |
| RBSLJAJQOVYTRQ-UHFFFAOYSA-N | |
| 546874 | |
| C1(=C(C(=O)C(=O)C1=O)O)O |
| C5H2O5 | |
| MFCD00181389 | |
| croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # | |
| 4,5-dihydroxycyclopent-4-ene-1,2,3-trione |
Specifications
| 488-86-8 | |
| Odorless | |
| MFCD00181389 | |
| croconic acid, crocic acid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione, unii-6hgy35uxaa, 6hgy35uxaa, 4-cyclopentene-1,2,3-trione, 4,5-dihydroxy, croconicacid, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione # | |
| RBSLJAJQOVYTRQ-UHFFFAOYSA-N | |
| 4,5-dihydroxycyclopent-4-ene-1,2,3-trione | |
| 546874 | |
| 98% |
| ∼280°C (decomposition) | |
| C5H2O5 | |
| 1 g | |
| Soluble in water and ethanol. | |
| C1(=C(C(=O)C(=O)C1=O)O)O | |
| 142.066 | |
| 142.07 | |
| Croconic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only