missing translation for 'onlineSavingsMsg'
Learn More

Crocein Orange G, TCI America™

Catalog No. C095125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C095125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C095125G Supplier TCI America Supplier No. C095125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1934-20-9
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.324
MDL Number MFCD00003912
InChI Key MWRMYQCXIXUBGJ-UHFFFAOYSA-M
Synonym acid orange 12, 2-naphthalenesulfonic acid, 6-hydroxy-5-2-phenyldiazenyl-, sodium salt 1:1, sodium 6-hydroxy-5-2-phenyldiazen-1-yl naphthalene-2-sulfonate, sodium 6-hydroxy-5-e-2-phenyldiazen-1-yl naphthalene-2-sulfonate
PubChem CID 23722701
IUPAC Name sodium;6-oxo-5-(phenylhydrazinylidene)naphthalene-2-sulfonate
SMILES C1=CC=C(C=C1)NN=C2C3=C(C=CC2=O)C=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 25 g
Formula Weight 350.32
Physical Form Crystalline Powder
Chemical Name or Material Crocein Orange G
RTECSNumber QK2430000
TSCA Yes