missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CrkL Recombinant Protein Antigen

Catalog No. NBP55130PEP
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP55130PEP 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP55130PEP Supplier R&D Systems™ Supplier No. NBP255130PEP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Novus product, part of R&D Systems. Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CRKL. The CrkL Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

Specifications

Gene ID (Entrez) 1399
Species Human
Product Line Novus
Protein Family CrkL
Purity >80% by SDS-PAGE and Coomassie blue staining
Content And Storage Store at -20 C. Avoid freeze-thaw cycles.
Formulation PBS and 1 M Urea, pH 7.4.
For Use With (Application) Antibody Competition
Gene Symbol CRKL
Molecular Weight (g/mol) M.W. (theoretical): 37 kDa
Quantity 100 μL
Regulatory Status RUO
Research Category Cancer, Phospho Specific, Signal Transduction
Source E. coli
Specific Reactivity This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-54145. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Show More Show Less