missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ CrkL Peptide
Specifications
Specifications
| Gene ID (Entrez) | 1399 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB300-884 |
| Gene Alias | crk-like protein, pCRKL, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like |
| Gene Symbol | CRKL |
| Quantity | 0.1 mg |
| Research Category | Cancer, Phospho Specific, Signal Transduction |