missing translation for 'onlineSavingsMsg'
Learn More

AlphaProTech Critical Cover™ Microbreathe™ Coveralls

Catalog No. 19811657
Change view
Click to view available options
Size:
3X-Large
5X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19811657 3X-Large
19811659 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19811657 Supplier AlphaProTech Supplier No. CV 04632 6TH
Only null left

Strong, flexible Microbreathe fabric coveralls provide outstanding protection, breathability, and superior barrier properties in controlled environments.

  • Barrier to particles, biologicals and fluids
  • Excellent vapor transmission rate (breatheability)
  • High filtration efficiency with low particle levels
  • Static dissipative

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Thumb Loop
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Back Style Elastic
Description Elastic wrist, ankle and back, Thumb loop
For Use With (Application) For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency
Style MicroBreathe™
Show More Show Less