missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AlphaProTech Critical Cover™ GenPro™ Lab Coats
Soft polypropylene lab coats designed for non-hazardous environments.
Supplier: AlphaProTech LC124212
Description
- Designed for non-hazardous, general purpose environments
- Ideal for personnel as well as facility visitors
- Offer comfort and breathability
- Full length, no-pocket design helps you control contaminants from entering or leaving sensitive areas
Specifications
| Controlled Environments Lab Coat | |
| White | |
| Single-use | |
| None | |
| Serged | |
| 100.3 cm |
| Polypropylene | |
| Snaps | |
| Non-sterile | |
| Tapered | |
| 39.5 in. | |
| 76.8 cm |