Learn More
Invitrogen™ CRISPR TrueGuide™ on-line tool
TrueGuide Synthetic gRNAs are ready-to-use, synthetic gRNAs that help enable researchers to truly harness the power of genome editing.
Supplier: Invitrogen™ A35533
Description
Invitrogen TrueGuide Synthetic gRNAs are ready-to-use, synthetic gRNAs designed to provide specific and high-efficiency knockout of your target gene.
TrueGuide Synthetic gRNAs both maximize and simplify the performance of genome editing experiments so researchers can create more relevant cellular models and make new discoveries. Custom TrueGuide Synthetic gRNAs are available in two package sizes: 1.5 nmol and 3 nmol.
Specifications
| Synthetic gRNA | |
| 1 Dried RNA Oligo, Store at Room Temperature | |
| 1.5 nmol | |
| TrueGuide |
For Research Use Only. Not for use in diagnostic procedures.