missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Crimp seals with PTFE/Rubber Septa Red Aluminum Seal

Catalog No. 111009837
Change view
Click to view available options
No. per Pack:
1000
Material:
Aluminum (Seal), PTFE
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack Material
111009837 1000 Aluminum (Seal), PTFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111009837 Supplier MilliporeSigma™ Supelco™ Supplier No. 27473

Closure type crimp seal

  • The PTFE/rubber septa are compatible with most organic solvents until pierced and are not recommended for aromatics, carbon disulfides, chlorinated solvents, hydrocarbons.

Specifications

Diameter (Metric) Inner 11 mm
Color Red
Material Aluminum (Seal), PTFE
Septum Rubber
No. per Pack 1000
For Use With (Equipment) 11 mm Crimp Top Vial