missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cresyl Violet acetate, pure, high purity biological stain

CAS: 10510-54-0 | C18H15N3O3 | 321.34 g/mol

$206.77 - $684.64

Chemical Identifiers

CAS 10510-54-0
Molecular Formula C18H15N3O3
Molecular Weight (g/mol) 321.34
MDL Number MFCD00013151,MFCD00013151
InChI Key XKOCOMKJPWEOHX-UHFFFAOYSA-M
PubChem CID 44134641
IUPAC Name acetic acid;5-iminobenzo[a]phenoxazin-9-amine
SMILES CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC229630050
SDS View Documents
Thermo Scientific Chemicals
229630050
5 g
Each for $206.77
Only null left
 
AC229630250
SDS View Documents
Thermo Scientific Chemicals
229630250
25 g
Each for $684.64
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10510-54-0
321.34
XKOCOMKJPWEOHX-UHFFFAOYSA-M
acetic acid;5-iminobenzo[a]phenoxazin-9-amine
C18H15N3O3
MFCD00013151,MFCD00013151
44134641
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3

Specifications

140.0°C to 143.0°C
65% min.
MFCD00013151,MFCD00013151
XKOCOMKJPWEOHX-UHFFFAOYSA-M
CC([O-])=O.NC1=CC2=[O+]C3=C(N=C2C=C1)C1=CC=CC=C1C(N)=C3
321.34
20% max. (50°C) (0.05mm Hg)
Pure
Black to Green
Fine Crystalline Powder
10510-54-0
C18H15N3O3
Solubility in water: soluble in water. Other solubilities: soluble in ethanol, insoluble in aceton
Authentic
acetic acid;5-iminobenzo[a]phenoxazin-9-amine
44134641
321.34
Glass bottle
5 g
Cresyl violet acetate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

RUO – Research Use Only