missing translation for 'onlineSavingsMsg'
Learn More

Cresol, TCI America™

Catalog No. C041225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C041225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C041225G Supplier TCI America Supplier No. C041225G
Only null left

Chemical Identifiers

CAS 1319-77-3
Molecular Formula C7H8O
MDL Number MFCD00151099
Synonym Cresylic Acid, Tricresol

Specifications

Color Yellow
Boiling Point 192°C
Quantity 25 g
UN Number 2022
Formula Weight 108.14
Physical Form Liquid
Chemical Name or Material Cresol
EINECSNumber (3)-0499&(4)-0057
RTECSNumber GO5950000
TSCA Yes