Learn More
Thermo Scientific Chemicals Cresol Red
Description
Cresol red is commonly used with lead acetate for the detection of urease after PAGE. It is also used as an indicator with the following transition intervals: pH 1.8 (orange) to pH 2.0 (yellow); pH 7.0 (yellow) to pH 8.8 (violet). Cresol Red is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approx. 125 bp DNA. Cresol Red does not inhibit Taq polymerase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cresol red is commonly used with lead acetate for the detection of urease after PAGE. It is also used as an indicator with the following transition intervals: pH 1.8 (orange) to pH 2.0 (yellow); pH 7.0 (yellow) to pH 8.8 (violet). Cresol Red is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) electrophoresis. In agarose, Cresol Red runs with an apparent molecular size of approx. 125 bp DNA. Cresol Red does not inhibit Taq polymerase.
Solubility
Soluble in water (slightly at 20°C), 50% ethanol (0.1%), dil. acids, and dil. alkalis. Insoluble in benzene.
Notes
Transition interval (acid): pH 1.8 (orange) to pH 2.0 (yellow); (alkaline) pH 7.0 (yellow) to pH 8.8 (violet)
Specifications
Specifications
| Melting Point | >300°C |
| Beilstein | 343399 |
| Merck Index | 14,2578 |
| Solubility Information | Soluble in water (slightly at 20°C),50% ethanol (0.1%),dil. acids,and dil. alkalis. Insoluble in benzene. |
| Quantity | 100 g |
| Formula Weight | 382.44 |
| Chemical Name or Material | Cresol Red |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| EINECSNumber | 217-064-2 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only